Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accessories

Supplier: GE Healthcare




 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Tricorn™ packing connector 5-5 1 18-1153-21
Tricorn™ packing connector 5-10 1 18-1153-22
Tricorn™ packing connector 10-10 1 18-1153-23
Tricorn™ packing equipment 5/50 1 18-1153-24
Tricorn™ packing equipment 10/100 1 18-1153-25
Tricorn™ glass tube 5/20 1 18-1153-04
Tricorn™ glass tube 5/50 1 18-1153-05
Tricorn™ glass tube 5/100 1 18-1153-06
Tricorn™ glass tube 5/150 1 18-1153-07
Tricorn™ glass tube 5/200 1 18-1153-08
Tricorn™ glass tube 10/20 1 18-1153-13
Tricorn™ glass tube 10/50 1 18-1153-14
Tricorn™ glass tube 10/100 1 18-1153-15
Tricorn™ glass tube 10/150 1 18-1153-16
Tricorn™ glass tube 10/200 1 18-1153-17
Tricorn™ glass tube 10/300 1 18-1153-18
Tricorn™ glass tube 10/600 1 18-1153-19
Tricorn™ 5 adapter unit 1 28-4064-06
Tricorn™ 5 bottom unit 1 18-1153-01
Tricorn™ 10 adapter unit 1 28-4064-07
Tricorn™ 10 bottom unit 1 18-1153-10
Tricorn™ 5 coarse filter kit 1 KIT 11-0012-53
Tricorn™ 10 coarse filter kit 1 KIT 11-0012-54
Filter tool 1 18-1153-20
Tricorn™ O-ring packing kit 1 KIT 18-1152-82
Tricorn™ O-ring 3×1 mm 2 18-1153-03
Tricorn™ O-ring 7×1 mm 2 18-1153-12
Tricorn™ O-ring 12×1 mm 2 18-1153-26
PEEK tubing 1/16", inner Ø 0,5 mm 2 m 18-1113-68
PEEK tubing 1/16", inner Ø 0,75 mm 1 18-1112-53
Fingertight connector 1/16" male 10 18-1112-55
Tricorn™ 10 filter kit, 7 µm porosity 1 KIT GEHE29-0536-12
Tricorn™ 5 filter kit, 7 µm porosity 1 KIT GEHE29-0535-86
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty