Methylcyclopentadienylmanganese tricarbonyl

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

EHERC15085036EA 0 GBP
EHERC15085036
Methylcyclopentadienylmanganese tricarbonyl
Methylcyclopentadienylmanganese tricarbonyl
Formula: C₉H₇MnO₃
MW: 218.09 g/mol
MDL Number: MFCD00001426
CAS Number: 12108-13-3
EINECS: 235-166-5
UN: 3281
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR