Would you like to log in again?
Sets of cutlery, sampling tools
Supplier: VWR Collection
Made from high impact PS. Disposable tool for sampling and sensorial tests made of highly resistant plastic, irradiated and individually packed in bag with leakproof closure to avoid contamination. Snap re-closing allows the bag to be used as a container for safe transfer and to transport samples.
  • Packed ready for single use and disposal
  • Easy to open bag closing system
  • Lot number identification for traceability according to GLP and GMP
  • Irradiated by beta radiation
  • Individually packed
Suitable for bacteriological and chemical analysis, ideal for use in food, dairy, pharmaceutical, cosmetic and chemical sectors.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Fork 100 481-0676
Knife 100 481-0675
Spoon 100 481-0674
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty