Thiazole-2-carbaldehyde
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Thiazolecarboxaldehyde
, Thiazole-2-carboxaldehyde
MOLEM61324267EA
0
GBP
MOLEM61324267
Thiazole-2-carbaldehyde
Thiazole-2-carbaldehyde
Formula:
C₄H₃NOS MW: 113.13 g/mol |
MDL Number:
MFCD00142924 CAS Number: 10200-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...