β-Nitrostyrene for synthesis, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(1-Nitro-2-phenyl)ethylene
, Nitrostyrene
, (2-Nitrovinyl)benzene
, ω-Nitrostyrene
M-Clarity™ quality level = MQ200
8.18165.0025EA
48
GBP
8.18165.9999
8.18165.0025
β-Nitrostyrene for synthesis, Sigma-Aldrich®
β-Nitrostyrene
Formula:
C₈H₇NO₂ MW: 149.14 g/mol Storage Temperature: Ambient |
CAS Number:
5153-67-3 EINECS: 203-066-0 UN: 2811 ADR: 6.1,II |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 54 - 57 °C |
Molar Mass | 149.14 g/mol |
Boiling Point | 250 - 260 °C |
Ignition Point | 250 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...