5-Hydroxy-1,4-naphthoquinone (juglone)

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Hydroxy-1,4-naphthalenedione5-Hydroxy-p-naphthoquinone

ROTH5843.2EA 0 GBP
ROTH5843.2
5-Hydroxy-1,4-naphthoquinone (juglone)
5-Hydroxy-1,4-naphthoquinone (Juglone)
Formula: C₁₀H₆O₃
MW: 174.16 g/mol
MDL Number: MFCD00001684
CAS Number: 481-39-0
EINECS: 207-567-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR