Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Power supply, PS200HC

Supplier: Hoefer
The PS200HC 200 V high current power supply is ideal for large electrophoretic blotting applications and for low voltage electrophoresis runs. The small footprint, large handle, and simple operation makes the PS200HC easy to set up and use.

  • Constant voltage or constant current mode with automatic crossover
  • 5 - 200 V, 10 - 2000 mA, 200 W
  • 4 sets of 4 mm output jacks
  • Continuous runs or 999 minute timer
  • Automatic recovery after power failure
  • Open circuit detection
  • Short circuit protection



 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
PS200HC, high current power supply, 200 V 1 732-1693
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Model PS200HC
Output voltage range/increments (V) 5 - 200/1
Input voltage (V) 100 - 240 VAC, 50/60 Hz
Current (mA) 10 - 2000
Timer 0 - 999 minutes
Display LED
Operating constant modes Current, voltage
Relative humidity (%) 80
W×D×H (mm) 125×280×335
Weight (kg) 2,4
Output 4 in parallel, 4 mm sockets

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk VWR Catalog Number Quantity
Power supply adapter, 4 mm to 4 mm, adapts old style output jacks to accept safety HV leads 1 KIT HOEFPSA410
Power cord, detachable, 230 VAC 1 HOEFPSCORD-230V
Fuse 0,25 A/250 V SB 3AG 5 HOEFPSF.25A-SB-3AG
Fuse 0,4 A/250 V, SB 5 HOEFPSF.4A-SB-5X20
Fuse 0,8 A, 250 V, SB 5 HOEFPSF.8A-SB-5X20
Fuse, 1,6 A, 250 V, microfuse 5 HOEFPSF1.6A-MICRO
Fuse, 1 A, 250 V 5 HOEFPSF1A-SB-3AG
Fuse, 1 A/250 V SB 15×20 5 HOEFPSF1A-SB-5X20
Fuse, 2,5 A/250 V SB 3AG 5 HOEFPSF2.5A-SB-3AG
Fuse, 2 A, 250 V, 5×20 mm, SB 5 HOEFPSF2A-SB-5X20
Fuse, 3 A, 250 V, 5×20 mm, SB 5 HOEFPSF3A-SB-5X20
Power supply adapter, 2 mm to 4 mm 1 KIT HOEFPSA230
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty