Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Block heaters, analogue, Thermubloc, BH-T series

Supplier: NICKEL ELECTRO
Clifton® block heaters provide a safe, dry and consistent method of heating accessory aluminium blocks with excellent thermal conductivity for heating tubes. Features a thermal over temperature protection, chemical resistant control panel, corrosion resistent stainless steel case and anti-bacterial technology.

  • The paint finish actively inhibits bacterial growth improving hygiene requirements as demanded by many clinical and laboratory environments
  • This anti-bacterial agent is integral within the paint finish




 

ORDER

Expand All / Collapse All
Type No. of blocks Temp. range Temp. stability Heating capacity W×D×H Weight Pk VWR Catalog Number Quantity
BH-1T 2* RT* +5...120 °C ±0,5 °C 0,25 kW 200×295×85 mm 2,8 1 NICKBH-1T

Blocks
Description Well size For No. of holes Depth W×D×H Pk VWR Catalog Number Quantity
Single block, plain - Microscope slides - - 140×50×72 mm 1 NICKDBK-0
Two blocks - 0,2 ml micro tubes or 6×8 PCR strips 54 - 140×50×72 mm 1 NICKDBK-0.2
Two blocks Ø 8 mm 0,5 ml micro tubes and PCR strips 16 28 mm 140×50×72 mm 1 NICKDBK-0.5
Two blocks Ø 11 mm 1,5 ml micro tubes 12 38 mm 140×50×72 mm 1 NICKDBK-1.5
Two blocks - 2,0 ml micro tubes (U well bottom) and PCR strips 16 - 140×50×72 mm 1 NICKDBK-2.0
Single block Ø 10 mm 10 mm Ø tubes (round bottom) 32 41 mm 138×74×45 mm 1 NICKDBK-10
Single block Ø 12 mm 12 mm Ø tubes (round bottom) 32 41 mm 138×74×45 mm 1 NICKDBK-12
Single block Ø 13 mm 13 mm Ø tubes (round bottom) 32 41 mm 138×74×45 mm 1 NICKDBK-13
Single block Ø 16 mm 16 mm Ø tubes (round bottom) 18 41 mm 138×74×45 mm 1 NICKDBK-16
Single block Ø 19 mm 19 mm Ø tubes (round bottom) 10 41 mm 138×74×45 mm 1 NICKDBK-19
Single block Ø 25 mm 25 mm Ø tubes and universal bottles 10 - 138×74×45 mm 1 NICKDBK-25
Single block Ø 29 mm 50 ml microcentrifuge tubes or bottles 8 - 138×74×45 mm 1 NICKDBK-50
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk For VWR Catalog Number Quantity
Hinged safety cover lid, semi-transparent 1 Clifton BH range block heaters NICKBHL-1
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty