4-Chloro-6-methylquinoline
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBHC-2757-5GEA
0
GBP
COBBHC-2757-250M
COBBHC-2757-5G
COBBHC-2757-1G
4-Chloro-6-methylquinoline
4-Chloro-6-methylquinoline
Formula:
C₁₀H₈ClN MW: 177.63 g/mol |
MDL Number:
MFCD02684204 CAS Number: 18436-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...