Carvone
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone
EHERC11052000EA
128
GBP
EHERC11052000
Carvone
Carvone
Formula:
C₁₀H₁₄O MW: 150.2176 g/mol |
MDL Number:
MFCD00062996 CAS Number: 99-49-0 EINECS: 202-759-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...