Decafluorobenzhydrol ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
L02197.06EA
94.1
GBP
L02197.06
Decafluorobenzhydrol ≥97%
Decafluorobenzhydrol
Formula:
C₁₃H₂F₁₀O MW: 364.14 g/mol Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000297 CAS Number: 1766-76-3 EINECS: 217-185-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...