Dibutyl succinate
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Succinic acid dibutyl ester
EHERC16985200EA
243
GBP
EHERC16985200
Dibutyl succinate
Dibutyl succinate
Formula:
C₁₂H₂₂O₄ MW: 230.3 g/mol |
MDL Number:
MFCD00040495 CAS Number: 141-03-7 EINECS: 205-449-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...