Would you like to log in again?
PE gloves, Manuplast® PE
Supplier: B. Braun
These disposable PE gloves are suitable for non hazardous, non medical applications.
  • Non sterile, powder-free
  • Latex-free
  • Flat, welded seam
  • Embossed surface for good grip
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Size Colour Length Thickness Packed Pk VWR Catalog Number Quantity
S Transparent 285 mm 0,023 mm 100/dispenser pack 100 112-2126
L Transparent 300 mm 0,023 mm 100/dispenser pack 100 112-2127
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Ambidextrous/hand-specific Ambidextrous
Disposable/reusable Disposable
Latex/Latex-free Latex-free
Powdered/powder-free Powder-free
Finish Embossed
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty