Digital immersion refractometer, PAN-1
Supplier: Atago
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ATAG3596EA 1810 GBP
ATAG3596
Digital immersion refractometer, PAN-1
Automatic instrument for continuous measurements of the refractive index in a liquid.
  • Reliable long-term performance
  • Immersed part made of SUS 316 stainless steel
  • Can display the converted value in Brix as a percentage of sugar in water
The concentration of a substance other than sugar can be calculated by creating a conversion table of the desired concentration percentage and corresponding Brix percentage.
Delivery information: Supplied with an alkaline battery compatible with NiMH.