Vortex mixers, pulsating, Vortex-Genie® Pulse
Supplier: Scientific Industries
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Genie®
444-0954EA 942 GBP
444-0954 444-0955 SCIISI-P276
Vortex mixers, pulsating, Vortex-Genie® Pulse
Shakers Vortex Shakers
Rugged and reliable vortex mixers with solid metal casing and rubber feet that prevent them 'walking'. Vortex-Genie® Pulse is ideal for applications requiring more aggressive action than standard vortexing.
  • The true pulsing action gives more collisions and random motion
  • Variable 'RUN/STOP' time provides quick pulsing or interval mixing
  • Can also be used as a standard 'continuous' vortex mixer
  • Digital control and display of speed, time, and 'RUN/STOP' time, for reproducibility and accuracy, with true speed display
Delivery information: Vortex-Genie® Pulse is supplied with quick change standard pop-off cup for single test tubes and 76 mm platform suitable for beakers, flasks and multiple tests tubes. Unit can be used with other optional Vortex-Genie accessories, which must be purchased separately.