Proteinase K
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E195-5MLEA 122 GBP
VWRCE195-5ML
Proteinase K
A non specific serine protease with a molecular weight of approximately 18 kDa. This enzyme exhibits high stability and activity in the presence of SDS, EDTA and urea, as well as over a wide pH range. Activity >30 units/mg.
Proteinase K is useful for the inactivation of nucleases during the isolation of DNA and RNA. Active over a broad pH range.