Tris-glycine-SDS (TGS) buffer solution, 10X liquid concentrate, Ultra Pure Grade
Supplier: VWR Life Science
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0783-1LEA 59.8 GBP
0783-1L 0783-4L 0783-5L
Tris-glycine-SDS (TGS) buffer solution, 10X liquid concentrate, Ultra Pure Grade
Buffers Electrophoresis Buffers Running Buffers
Liquid concentrate used for protein electrophoresis to maintain pH and temperature of the environment as well as provide ions that carry electrical current.