Glycerine ≥99.0%, Proteomics Grade, sterile
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Glycerol
, 1,2,3-Propanetriol
M153-100MLEA
48.6
GBP
M153-100ML
Glycerine ≥99.0%, Proteomics Grade, sterile
Glycerine
Formula:
HOCH₂CH(OH)CH₂OH MW: 92.09 g/mol Boiling Pt: 182 °C Melting Pt: 20 °C Density: 1.26 g/cm³ (20 °C) Flash Pt: 177 °C (open cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
![]() |
Specification Test Results
Arsenic |
|
Ash | 0.01% |
DNase | None |
Heavy metals (as Pb) |
|
Protease | None |
Purity (Anhydrous) | 99.0% |
RNase | None |
Specific gravity | 1.250 − 1.265 |
Sterility | Pass |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...