Glycerine ≥99% for biotechnology, sterile

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Glycerol1,2,3-Propanetriol

E520-100MLEA 84 GBP
E520-100ML
Glycerine ≥99% for biotechnology, sterile
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18.6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C (open cup)
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://uk.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg

Order Now

Specification Test Results

Arsenic 0.0003%
DNase none detected
Heavy Metals (as Pb) 0.0005%
Purity (Anhydrous) 99%
Residue on Ignition 0.01%
RNase none detected
Specific Gravity 1.2615 – 1.2635
Sterility Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR