Tool kit
Supplier: RSG Solingen
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
238-0025EA 725 GBP
RSGA000.200
Tool kit
High quality tool kit ideal for use in pharma or food laboratories. All parts are made from high-grade stainless steel, which prevents corrosion when installing components also made from stainless steel.
  • Uncoated, preventing any possible allergic reaction
  • Static load of the ratchet is 30% higher than that of a standard model
  • Maximum use of the screwdriver inserts are 3 times higher