Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

General sequencing accessories

Supplier: C.B.S. Scientific




 

ORDER

Expand All / Collapse All

Bar clamps
Description Pk VWR Catalog Number Quantity
Bar clamp for 20 cm wide sequencer 1 CBSCGPC-1000
Bar clamp for 33 cm wide sequencer 1 CBSCGPC-2000

Gel Slider™ (sliding gel caster)
Description Pk VWR Catalog Number Quantity
Gel Slider™, casts glass plates 20 - 38 cm (w)×22 - 62 cm (l) 1 CBSCGSL-2001
Drip tray for 20 cm wide glass plates 1 CBSCDT-20
Drip tray for 33 cm wide glass plates 1 CBSCDT-33

Gel sealing tape
Description Pk VWR Catalog Number Quantity
Gel sealing tape, 25 mm×200 m 1 Roll 730-1632
Gel sealing tape, 38 mm×200 m 1 730-1633

Glass plate rack
Description Pk VWR Catalog Number Quantity
Glass plate rack, black, 12-place 1 CBSCGRH-100B
Mini-Glass plate rack, black, 20-place 1 CBSCGR-200-20B

Rubber ears
Description Pk VWR Catalog Number Quantity
Large white clamp for holding thicker sandwiches together (clamps glass pates to upper reservoir, holds aluminium in place) 6 730-0983
White clamp, large, for holding thinner sandwiches together (for gel casting and holding glass plates together, without aluminium) 6 730-0984

White clamps
Description Pk VWR Catalog Number Quantity
Gel thermometer strip, adhesive 1 CBSCEGT-100
Gel thermometer strip, adhesive 10 CBSCEGT-100-10
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty