Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Repetitive pipette, Minipet®

Supplier: Bel-Art Products
Minipet®
Minipet® pipette offers a high level of chemical resistance and may be used with wirtually any solvent or acid (except HF). Fluids come in contact with only borosilicate glass and PTFE.

  • Three-way check valve for continuous, repetitive pipetting
  • Molded PP handle offers comfort for extended use
  • Turning the knob on top sets the self-locking capacity to ensure dispensing repeatibility of ±0,5%
  • Autoclavable

Ordering information: Supplied with a chrome plated check valve, Luer lock fitting, Buna-N-seals, stainless steel spring, and Luer lock canula (457 mm long, 3,2 mm ID), silicone filling tube and PTFE sinker.



 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Minipet® pipettor, 1 ml, PTFE 1 612-6040
Minipet® pipettor, 10 ml, PTFE 1 612-6042
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Volume (µl) 1000 10000

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty