Dipotassium phthalate for synthesis, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
8.22080.0100EA
65.1
GBP
8.22080.9999
8.22080.9010
8.22080.0100
Dipotassium phthalate for synthesis, Sigma-Aldrich®
Dipotassium phthalate
Formula:
C₈H₄K₂O₄ MW: 242.31 g/mol |
CAS Number:
4409-98-7 EINECS: 224-561-8 |
Specification Test Results
Solubility | (20 °C) soluble |
Molar Mass | 242.32 g/mol |
Hill Formula | C8H4K2O4 |
Chemical Formula | C6H4-1,2-(COOK)2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...