Propachlor, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

45637-250MGEA 60.5 GBP
45637-250MG
Propachlor, Supelco®
Propachlor
Formula: C₁₁H₁₄ClNO
MW: 211.69 g/mol
CAS Number: 1918-16-7
EINECS: 217-638-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR