Diethyl disulphide for synthesis, Sigma-Aldrich®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
8.43957.0250EA
0
GBP
8.43957.0250
Diethyl disulphide for synthesis, Sigma-Aldrich®
Diethyl disulphide
Formula:
C₄H₁₀S₂ MW: 122.26 g/mol Boiling Pt: 152 °C (1013 hPa) Melting Pt: –101.5 °C Density: 0.984 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009266 CAS Number: 110-81-6 EINECS: 203-805-7 UN: 1993 ADR: 3,III |
Specification Test Results
Solubility | 0.3 g/l (25°C) |
Vapor Pressure | 5.7 hPa (25 °C) |
Flash Point | 40 °C |
Density | 0.993 g/cm3 (25 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...