Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Büchner funnels, table-top with porous filter or perforated plate

Supplier: Bel-Art
SCIENCEWARE®
PE, with Fritware® porous filter or HDPE perforated plate.

Fixed or removable Fritware® porous filter plates are supported by a multiple ring grid. They are made of HDPE, 6,4 mm thick, with a non-porous ring at the periphery of the plate which seals filter paper when used. Available in two porosities (medium and coarse), the medium porosity plate is 45 - 90 µm while the coarse porosity plate is 90 - 130 µm.

Fixed or removable perforated filter plates are made of 4,8 mm thick HDPE with 4,8 mm perforations on 11 mm centers. The fixed version is for coarse filtration or use with a cloth or paper filter. The removable filter plate is available for use in precious metals recovery.

Ordering information: Removable and medium porosity filter plates are also available; contact your VWR sales office for more information.

Caution: Safe to use at the temperature range from –80 to 52 °C.



 

ORDER

Expand All / Collapse All

Funnels with fixed plate
Description For filter Ø Plate Ø Pk VWR Catalog Number Quantity
Medium porosity plate 240 mm 260 mm 1 BELAH146253510
Perforated plate 240 mm 260 mm 1 BELAH146270000
Medium porosity plate 420 mm 457 mm 1 BELAH146253518
Medium porosity plate 500 mm 600 mm 1 BELAH146253524
Medium porosity plate 900 mm 914 mm 1 BELAH146253536

Funnels with removable plate
Description For filter Ø Plate Ø Pk VWR Catalog Number Quantity
Medium porosity plate 240 mm 260 mm 1 BELAH146263510
Perforated plate 240 mm 260 mm 1 BELAH146271260
Coarse porosity plate 240 mm 260 mm 1 BELAH146201260
Perforated plate 420 mm 457 mm 1 BELAH146271457
Medium porosity plate 420 mm 457 mm 1 BELAH146263518
Coarse porosity plate 420 mm 457 mm 1 BELAH146211457
Perforated plate 500 mm 610 mm 1 BELAH146271610
Medium porosity plate 500 mm 610 mm 1 BELAH146263524
Coarse porosity plate 500 mm 610 mm 1 BELAH146221610
Medium porosity plate 900 mm 914 mm 1 BELAH146263536
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping