Contact plates for isolators and cleanrooms, Envirocheck® Contact
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.18408.0020EA 44.7 GBP
MERC1.18408.0020
Contact plates for isolators and cleanrooms, Envirocheck® Contact
As a simple and efficient way to detect microbiological contamination, Envirocheck® Contact plates are used to screen for bacteria, yeasts and moulds related to contamination in the production process. The contact plates serve to examine surfaces in isolators and/or cleanrooms. In order to fulfil the requirements for protected areas, all Envirocheck® Contact plates for isolators and cleanrooms are triple wrapped and gamma irradiated. The inner bag is impermeable to H₂O₂ and includes a pouch containing a drying agent to minimise the formation of condensation. To inactivate any residual disinfectants on the surface to be tested, the Envirocheck® Contact plates for isolators and cleanrooms additionally contain 4 neutralising agents. All plates are stored at room temperature (15 to 25 °C), hence refrigeration is not required. Storage at site of application is possible.
  • Gamma irradiated, triple wrapped, H₂O₂-impermeable inner bag
  • Four neutralisers to inactivate any residual disinfectants
  • A pouch containing a drying agent to minimise the formation of condensation
  • Long shelf life - up to 9 months and long incubation time possible
  • Product name, expiry date, lot number and serial production number are imprinted on each plate for safe and easy product identification and shelf life monitorinG
Packaging: Triple wrapped, 2 sets of 10 plates each.