Would you like to log in again?
Laboratory installation SafetyWasteCaps, collector for HPLC waste
Supplier: SCAT Europe
SafetyWasteCaps conduct the waste tubes of your HPLC system to the disposal system, directly and air-tight. Escape of harmful solvent vapours are effectively prevented.
All parts coming in contact with fluids are manufactured from pure PTFE, for optimum chemical resistance even against aggressive solvents and acids.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
SafetyWasteCap II, with 2 connectors and 6 capillary fittings (2× 1,6 mm OD, 2× 2,3 mm OD and 2× 3,2 mm OD) 1 SCTV106489
SafetyWasteCap II, with 3 connectors, 7 capillary fittings (2× 1,6 mm OD, 2× 2,3 mm OD and 2× 3,2 mm OD) and 1 stepped curved fitting (1× 6,4 - 9 mm OD) 1 SCTV106497
SafetyWasteCap III, with 3 connectors and 9 capillary fittings (3× 1,6 mm OD, 3× 2,3 mm OD and 3× 3,2 mm OD) 1 SCTV106491
SafetyWasteCap IV, with 4 connectors and 12 capillary fittings (4× 1,6 mm OD, 4× 2,3 mm OD and 4× 3,2 mm OD) 1 SCTV106493
SafetyWasteCap IV, with 5 connectors, 12 capillary fittings (4× 1,6 mm OD, 4× 2,3 mm OD and 4× 3,2 mm OD) and 1 stepped curved fitting (1× 6,4 - 9 mm OD) 1 SCTV106498
SafetyWasteCap V, with 5 connectors and 15 capillary fittings (5× 1,6 mm OD, 5× 2,3 mm OD and 5× 3,2 mm OD) 1 SCTV106494
SafetyWasteCap VII, with 7 connectors and 21 capillary fittings (7× 1,6 mm OD, 7× 2,3 mm OD and 7× 3,2 mm OD) 1 SCTV106492
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty