Ficoll® 400, molecular biology grade, VWR®
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
437093TEA 0 GBP
VWRC437093T
Ficoll® 400, molecular biology grade, VWR®
Ficoll® 400 is a neutral, hydrophilic polymer of sucrose, which readily dissolves in aqueous solutions.
Ficoll® is a trademark owned by GE Healthcare companies.