Infrared thermometer, Traceable®
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
620-2096EA 255 GBP
VWRI620-2096
Infrared thermometer, Traceable®
This infrared thermometer provides the widest possible range for accurately measuring different surface temperatures.
  • Field of view is 7:1
  • Adjustable emissivity from 0,2 to 1,0 in increments of 0,01
  • Data 'HOLD' function, maximum/minimum memory and 1 second sampling
Infrared sensor and K-type beaded probe.
Delivery information: Supplied with Traceable® certificate of calibration, K-type beaded probe, carrying case and 9 volt battery.