Ice pans, Magic Touch™
Supplier: Bel-Art
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BELAM188484101EA 192 GBP
BELAM188484101
Ice pans, Magic Touch™
These pans are ideal for ice and water baths as well as general organization and transport of samples.
  • Use with ice, dry ice, dry ice alcohol slurries, liquid nitrogen
  • Stackable, lightweight, non-sweating, leakproof design
  • Expanded Polymer Foam is chemical resistant
  • Impervious to moisture and odors, and unbreakable under normal use conditions