Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biological safety cabinet, class II, type A2, Purifier® Logic®

Supplier: Labconco
Biological safety cabinets are designed with Human Inclination Technology™ which provides personnel, product and environmental protection from hazardous particulates such as agents that require biosafety level 1, 2 or 3 containments.

These biological safety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional canopy connection. When canopy-connected, the biosafety cabinets may be used for applications involving minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research.

During operation, room air is drawn into the inlet grill. The air in the plenum beneath the work surface is a mixture of unfiltered room air and air that has just passed through the work area. This contaminated air is drawn by the blower through the back plenum of the cabinet, where approximately 70% of the air is recirculated through the supply HEPA filter and back over the work area. The balance of the contaminated air is discharged to the environment after passing through the exhaust HEPA filter.

It is applicable for working with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates.




 

ORDER


The main product you are looking for is no longer available for purchase. Accompanying products and services may be found elsewhere on the page.

SPECIFICATIONS

Model
Exhaust air volume (m³/h)
Lighting system (lux)
Noise level (dB(A))
Electrical connection
Power rating (kW)
Ext. W×D×H (mm)
Weight (kg)
Filters

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk W×D×H VWR Catalog Number Quantity
Turn table for Purifier cabinets 1 - 135-2001
4' IV bar kit, includes IV bar, mounting hardware and four hangers, bar supports intravenous solution bottles and bags 1 - LBCP3858601
6' IV bar kit, includes IV bar, mounting hardware and four hangers, bar supports intravenous solution bottles and bags 1 - LBCP3858603
Back draft damper adjust the biosafety cabinet's exhaust airflow 1 - LBCP3858800
Stainless steel shelf mounts to rear grille of Purifier Logic Biosafety Cabinet or PuriCare Procedure Station, provides 203×127 mm surface, 1 kg capacity 1 - LBCP3859000
4' Telescoping base stand with leveling feet and non-welded shelf 1 - 135-1999
Electric hydraulic lift base stands, nominal width of 101,6 mm, 79 kg 1 - mm LBCP3780104
Vacu-pass cord and cable portal 1 KIT - LBCP3402800
Service fixture kit, includes serrated hose tip valve with ball valve, quarter turn control handle, hardware and instructions for plumbing to services 1 KIT - LBCP3747500
Hydraulic lift base stands, nominal width of 101,6 mm, 75 kg 1 - mm LBCP3780201
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty