Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Microtube racks, Mega

Supplier: VWR Collection

  • For tubes of diameter 10 - 13 mm
  • Storage of high quantity of samples eliminates the need for multiple racks
  • Alphanumeric grid for quick location and retrieval of tubes
  • Sturdy, lightweight material for easy transport

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
For tubes No. of holes W×D×H Colour Pk VWR Catalog Number Quantity
0,2/0,5/1,5/2,0/5,0 ml 216 337×267×91 mm White 1 211-0364
0,2/0,5/1,5/2,0/5,0 ml 216 337×267×91 mm Blue 1 211-0363
0,2/0,5/1,5/2,0/5,0 ml 432 673×230×80 mm White 1 212-0661
0,2/0,5/1,5/2,0/5,0 ml 432 673×230×80 mm Blue 1 212-0660
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web. will publish your name as user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty