Rack for slide trays
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
631-0964EA 0 GBP
VWRI631-0964
Rack for slide trays
Sturdy rack designed to hold five slide trays.
  • Matching clasps area for identification labels
  • Can be placed in refrigerators
  • Specifically used for cold storage of samples
Suitable for applications in microbiology, histology and pathology.