Pump tubing, Tygon® MHSL 2001
Supplier: Ismatec
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Tygon®
ISMC06518-22EA 496 GBP
ISMC06518-22 ISMC96466-14 ISMCSC0818 ISMC06518-66 ISMCSC0806 ISMCSC0804A ISMC96465-28 ISMC06518-04 ISMCSC0808A ISMC06518-64 ISMC06518-14 ISMC06518-16 ISMC96466-42 ISMC06475-18 ISMC06518-08
Pump tubing, Tygon® MHSL 2001
Tubing Peristaltic Pump Tubing
The transparent, plasticiser-free tubing with superior pump-life. Eespecially designed for MEK and other aggressive solvents.
  • Plasticizer and oil-free
  • Smooth inner-bore
  • Low sorption maintains fluid and tube integrity
  • Does not impart anything into the pumping medium
  • No release of hazardous materials when properly incinerated
  • Complies with FDA standards
Temperature resistant from −73 to +57 °C
Hardness (Shore durometer A): 69
Density: 0,88