Carbon disulphide ≥99.9%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

22518.292EA 0 GBP
22518.292
Carbon disulphide ≥99.9%, AnalaR NORMAPUR® analytical reagent
Carbon disulphide
Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46.2 °C (1013 hPa)
Melting Pt: –111.6 °C
Density: 1.2632 g/cm³ (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
Merck Index: 12,01859
https://uk.vwr.com/stibo/web/std.lang.all/53/94/785394.jpg

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Acidity or alkalinity Max. 0.0004 meq/g
Colouration Max. 10 APHA
Density (20/4) 1.262 to 1.264
Evaporation residue Max. 10 ppm
Foreign sulphur derivatives (as H2S) Max. 1 ppm
Water Max. 100 ppm
SO3 (as SO2) Max. 10 ppm
SO4 (Sulphate) Max. 2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR