Carbon disulphide, EMPLURA®, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

1.02211.1000EA 190 GBP
1.02211.1000
Carbon disulphide, EMPLURA®, Supelco®
Carbon disulphide
Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46.2 °C (1013 hPa)
Melting Pt: –111.6 °C
Density: 1.2632 g/cm³ (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3 (6.1),I
Merck Index: 12,01859
https://uk.vwr.com/stibo/web/std.lang.all/53/94/785394.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR