Carbon disulphide, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
1.02210.1000EA
189
GBP
1.02210.1000
Carbon disulphide, Uvasol® for spectroscopy, Supelco®
Carbon disulphide
Formula:
CS₂ MW: 76.14 g/mol Boiling Pt: 46.2 °C (1013 hPa) Melting Pt: –111.6 °C Density: 1.2632 g/cm³ (20 °C) Flash Pt: –30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011321 CAS Number: 75-15-0 EINECS: 200-843-6 UN: 1131 ADR: 3 (6.1),I Merck Index: 12,01859 |
Specification Test Results
Solubility | 2.1 g/L (20 °C) |
Melting Point | -111.6 °C |
Molar Mass | 76.13 g/mol |
Boiling Point | 46.5 °C (1013 hPa) |
Vapor Pressure | 398 hPa (20 °C) |
Flash Point | -30 °C |
Explosion Limit | 1 - 60%(V) |
Density | 1.26 g/cm³ (20 °C) |
Ignition Point | 95 °C |
Evaporation Number | 1.8 |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...