Silica gel, Pore size: 60 Å 0.060-0.2 mm (70-230 mesh)

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

42725.18EA 0 GBP
42725.18
Silica gel, Pore size: 60 Å 0.060-0.2 mm (70-230 mesh)
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR