Nitric acid fuming 100%, EMSURE® Reag. Ph. Eur. for analysis, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ300

1.00455.1000EA 210 GBP
1.00455.1000
Nitric acid fuming 100%, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 83 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
EINECS: 231-714-2
UN: 2032
ADR: 8 (5.1, 6.1),I
REACH: 01-2119487297-23
Merck Index: 12,06672

Order Now

Specification Test Results

Assay (acidimetric) ≥ 98.5 %
Identity passes test
Dilution test passes test
Chloride (Cl) ≤ 10 ppm
Heavy metals (as Pb) ≤ 5 ppm
Sulfate (SO₄) ≤ 300 ppm
Ag (Silver) ≤ 0.02 ppm
Al (Aluminium) ≤ 0.20 ppm
As (Arsenic) ≤ 0.01 ppm
Ba (Barium) ≤ 0.02 ppm
Be (Beryllium) ≤ 0.02 ppm
Bi (Bismuth) ≤ 0.10 ppm
Ca (Calcium) ≤ 0.15 ppm
Cd (Cadmium) ≤ 0.02 ppm
Co (Cobalt) ≤ 0.02 ppm
Cr (Chromium) ≤ 0.20 ppm
Cu (Copper) ≤ 0.02 ppm
Fe (Iron) ≤ 2.0 ppm
Ge (Germanium) ≤ 0.05 ppm
K (Potassium) ≤ 0.10 ppm
Li (Lithium) ≤ 0.02 ppm
Mg (Magnesium) ≤ 0.05 ppm
Mn (Manganese) ≤ 0.02 ppm
Mo (Molybdenum) ≤ 0.02 ppm
Na (Sodium) ≤ 0.50 ppm
Ni (Nickel) ≤ 0.10 ppm
Pb (Lead) ≤ 0.02 ppm
Sr (Strontium) ≤ 0.02 ppm
Ti (Titanium) ≤ 0.10 ppm
Tl (Thallium) ≤ 0.05 ppm
V (Vanadium) ≤ 0.02 ppm
Zn (Zinc) ≤ 0.05 ppm
Zr (Zirconium) ≤ 0.10 ppm
Residue on ignition (as SO₄) ≤ 50 ppm
According to the chemical properties of Nitric acid 100 % the influence of light can cause formation of nitrogen oxides.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR