Sodium chloride ≥99.5%, Electran for molecular biology
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Halite
, Sea salt
, Saline
443824TEA
36
GBP
443824T
Sodium chloride ≥99.5%, Electran for molecular biology
Sodium chloride
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 °C (1013 hPa) Melting Pt: 801 °C Density: 2.16 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Specification Test Results
Assay | Min. 99.5 % |
Appearance | White crystalline powder |
DNases (exo- and endonucleases) | Not detected |
RNases | Not detected |
Proteases | Not detected |
pH (5 %) | 5.0 - 8.0 |
Absorbance (260 nm) (0.1 mol/l) | Max. 0.01 |
Absorbance (280 nm) (0.1 mol/l) | 0.01 |
Heavy metals (as Pb) | Max. 0.001 % |
PO4 (Phosphate) | Max. 0.001 % |
SO4 (Sulphate) | Max. 0.01 % |
Ba (Barium) | Max. 0.001 % |
Br (Bromide) | Max. 0.005 % |
Ca (Calcium) | Max. 0.002 % |
Fe (Iron) | Max. 0.0001 % |
I (Iodide) | Max. 0.001 % |
K (Potassium) | Max. 0.005 % |
Mg (Magnesium) | Max. 0.001 % |
Pb (Lead) | Max. 0.0005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...