Sodium bromate, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Danger

71325-250GEA 59.8 GBP
71325-250G
Sodium bromate, Supelco®
Sodium bromate
Formula: BrNaO₃
MW: 150.89 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 381 °C
Density: 3.254 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003476
CAS Number: 7789-38-0
EINECS: 232-160-4
UN: 1494
ADR: 5.1,II
Merck Index: 12,08736

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR