Sodium tetrahydroborate ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Sodium tetrahydridoborateSodium borohydride

27885.180EA 60.5 GBP
27885.180 27885.134
Sodium tetrahydroborate ≥98%, GPR RECTAPUR®
Sodium tetrahydroborate
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3,I
Merck Index: 12,08735

For production and scale-up, we can support you!

Ask for larger pack sizes or bulk quantities that are not available in this page by using the customization request form.

Discover our service

Special packs and customized solutions

Explore more

Order Now

Specification Test Results

Assay (Hypochlorite) Min. 98.0 %
Identification (ICP-OES) Passes test

For production and scale-up, we can support you!

Ask for larger pack sizes or bulk quantities that are not available in this page by using the customization request form.

Discover our service

Special packs and customized solutions

Explore more

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR