Toluene reference substance for gas chromatography, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Phenylmethane
, Methylbenzene
M-Clarity™ quality level = MQ100
1.09768.0005EA
263
GBP
1.09768.0005
Toluene reference substance for gas chromatography, Supelco®
Toluene
Formula:
C₆H₅CH₃ MW: 92.14 g/mol Boiling Pt: 110.6 °C (1013 hPa) Melting Pt: –95 °C Density: 0.867 g/cm³ (20 °C) Flash Pt: 4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008512 CAS Number: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3, REACH: 05-2114615130-69 Merck Index: 12,09667 |
Specification Test Results
Solubility | 0.52 g/l (20 °C) |
Melting Point | -95 °C |
Molar Mass | 92.14 g/mol |
Boiling Point | 110.6 °C (1013 hPa) |
Vapor Pressure | 29 hPa (20 °C) |
Flash Point | 4 °C |
Refractive Index | 1.4968 (20 °C) |
Explosion Limit | 1.2 - 8 %(V) |
Density | 0.87 g/cm3 (20 °C) |
pH | (H2O) not applicable |
Ignition Point | 535 °C |
Viscosity kinematic | 0.7 mm2/s (20 °C) |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...