Toluene, AnalaR NORMAPUR® analytical reagent
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Phenylmethane
, Methylbenzene
102845FEA
0
GBP
102845F
Toluene, AnalaR NORMAPUR® analytical reagent
Toluene
Formula:
C₆H₅CH₃ MW: 92.14 g/mol Boiling Pt: 110.6 °C (1013 hPa) Melting Pt: –95 °C Density: 0.867 g/cm³ (20 °C) Flash Pt: 4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008512 CAS Number: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3,II REACH: 05-2114615130-69 Merck Index: 12,09667 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...