4-Ethylanisole ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Warning
|
Synonyms:
1-Ethyl-4-methoxybenzene
A10391.09EA
28.5
GBP
A10391.09
A10391.18
4-Ethylanisole ≥98%
4-Ethylanisole
Purity: 98+%
CAS No.: 1515-95-3
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 79°(174°F)
Refractive Index: 1.5085
Boiling Point: 195°
Beilstein Reference: 774365
MDL No.: MFCD00015176
Formula:
C₉H₁₂O MW: 136.19 g/mol Boiling Pt: 195 °C Density: 0.962 Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015176 CAS Number: 1515-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...