Sample tubes with handle, steriplast
Supplier: Bürkle
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BURK6206-0101EA 5.6 GBP
BURK6206-0101
Sample tubes with handle, steriplast
Polypropylene, blue transparent.
  • Sterile, packed separately
  • Suitable for use with foodstuffs
  • Very good chemical and temperature resistance
Sterile sampling tube with removable handle and tightly closing sealing cover. The lid can be opened with one hand and is then at a 90° angle. Liquid/condensate in the lid goes into an absorbing channel integrated into the lid, nothing is lost, nothing can drip.
The sterile sampling tube with a handle of 24 cm in length enables a clean sample to be placed directly in the sampling container. This mainly prevents cross-contamination.