ALFAA11011

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A11011.30EA 193.44 GBP
A11011.30 A11011.09 A11011.18
ALFAA11011
Purity: 99%
CAS No.: 104-20-1
Molecular Formula: C11H14O2
Formula Weight: 178.23
Flash Point: 110°(230°F)
Refractive Index: 1.5200
Melting Point: 7-9°
Boiling Point: 147-148°/8mm
Beilstein Reference: 1869592
MDL No.: MFCD00008791
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR