Phenol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Carbolic acid
, Hydroxybenzene
, Phenylalcohol
M-Clarity™ quality level = MQ200
SIAL16017-6X1KGEA
466
GBP
SIAL16017-6X1KG
SIAL16017-1KG
Phenol, Sigma-Aldrich®
Phenol
Formula:
C₆H₅OH MW: 94.11 g/mol Boiling Pt: 181.7 °C (1013 hPa) Melting Pt: 40.8 °C Density: 1.0722 g/cm³ (20 °C) Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002143 CAS Number: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1,II REACH: 01-2119471329-32 Merck Index: 12,07390 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...