3'-Bromo-4'-methoxybiphenyl-3-carboxylic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBCA-4948-1GEA 0 GBP
COBBCA-4948-1G
3'-Bromo-4'-methoxybiphenyl-3-carboxylic acid
3'-Bromo-4'-methoxybiphenyl-3-carboxylic acid
Formula: C₁₄H₁₁BrO₃
MDL Number: MFCD15143539
CAS Number: 1215206-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR